2-Hydroxy-4-(trifluoromethyl)pyridine 95%

Toimittaja: BLD PHARMATECH GMBH

BLDPBD9849-1GEA 18.7 EUR
BLDPBD9849-1G BLDPBD9849-5G BLDPBD9849-250MG BLDPBD9849-10G BLDPBD9849-100G BLDPBD9849-25G
2-Hydroxy-4-(trifluoromethyl)pyridine 95%
2-Hydroxy-4-(trifluoromethyl)pyridine

Storage condition: Inert atmosphere, Room Temperature

Kaava: C₆H₄F₃NO
MW: 163,1 g/mol
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00153195
CAS-numero: 50650-59-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR