5-Fluori-2-jodianiliini 97%

BLDPBD9914-1GEA 11.5 EUR
BLDPBD9914-1G BLDPBD9914-100G BLDPBD9914-5G BLDPBD9914-250MG BLDPBD9914-10G BLDPBD9914-25G
5-Fluori-2-jodianiliini 97%
5-Fluori-2-jodianiliini

Storage condition: Keep in dark place, Inert atmosphere, 2-8°C

Kaava: C₆H₅FIN
MW: 237,02 g/mol
Storage Temperature: Jääkaappi
MDL numero: MFCD02093953
CAS-numero: 255724-71-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR