N,N-Dimethylformamide-[carbonyl-¹³C] (99% ¹³C )

Toimittaja: LGC Standards PROMOCHEM
Vaara

PROOCIL-CLM-503-1EA 1060 EUR
PROOCIL-CLM-503-1
N,N-Dimethylformamide-[carbonyl-¹³C] (99% ¹³C )
N,N-Dimethylformamide-[carbonyl-¹³C]
Kaava: HCOON(CH₃)₂
CAS-numero: 32488-43-0
UN: 2265
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR