Centrifuge, Bench, Ventilated, SL 16R
Toimittaja: Thermo Scientific
Thermo Scientific™
HERA75004030EA 10560 EUR
HERA75004030
Centrifuge, Bench, Ventilated, SL 16R
Sentrifugit Pöytämalliset sentrifugit
Perform everyday sample processing with the Thermo Scientific™ SL 16 Centrifuge Series, which offers exceptional value, capacity and ergonomics for safe and simple operation. Additionally, proprietary innovations ensure safe, simple and comfortable daily operation in even the most rigorous environments.
  • Proprietary motorized lid latch enables one finger downward motion for easiest closin
  • Saves time switching between applications
  • Easy access to rotor chamber for quick cleaning
  • A healthier working environment and longer unit life
  • Biocontainment solution seals with a snap; eliminates screw caps and complicated clips
  • Glove-friendly
  • One-handed open/close capability
Intuitive controls simplify operation and provide detailed information on a wide range of operating parameters and processing status for optimal performance. Certified by CAMR™ in Porton Down, UK. Advanced rotor management system maximizes acceleration, braking and residual load imbalance for each rotor and bucket option, Optimizes safety and improves separations. Compact dimensions optimize work area and ergonomic height and curved design ensure easy sample loading and unloading and easy cleaning. Up to 40% energy consumption savings on industry standard protocols, such as standard blood separation or conical tubes processing. For separations at ambient temperatures, CFC-free refrigeration system.Temperature Range: −10 to +40 °C, Pre-Cooling Function with direct button.