Guide your students through basic identification of bullets and shell casings with real evidence samples and accurate re-created examples. The Flashcard Version of the activity uses flashcard images in place of activity materials for comparison. Students will determine the trajectory of a fired bullet, decipher the calibre of a bullet and shell casing, and learn techniques used to identify and match weapons to bullets and shell casings.
- Multiple techniques covered in this activity divided into 5 sections
- Time requirement: Approx 2 hours
Access to a dissection microscope would be useful but not a requirement. Please see elsewhere in the catalogue for details of microscopes.
Toimitustiedot: Includes materials for four groups, teacher’s guide, student copymasters.