Photo rules, foldable, Ward's®
818-0148EA 30.8 EUR
WARD154614
Photo rules, foldable, Ward's®
Precisely measure irregularly-shaped objects in two dimensions with these dual-sided, flexible plastic rules. Both sides of the rule pivot at a joint located on one end, allowing for easy storage. British Imperial and metric units are printed on opposite sides of the rule.
  • Graduated in ¼" and 0,5 cm increments