Hair and Fibre Analysis, Laboratory Activity Set
Toimittaja: Ward's
761-0333EA 218 EUR
WARD366146 WARD366145
Hair and Fibre Analysis, Laboratory Activity Set
Learn to determine the species of origin from a hair sample, differentiate between natural and synthetic fibres, and perform tests to identify the specific type of fibre with this engaging activity. Students will use their observational skills and a compound microscope (available separately) to examine the morphological characteristics of hair and fibre.
  • Time requirement: 2/3 x 45 minute laboratory periods
  • Refill kit available to replenish consumables
Toimitustiedot: The activity includes a human hair sample set, animal hair sample set, fibre sample set, as well as a teacher’s guide, student copymasters, and materials for 15 groups.