Osteometric board
Toimittaja: WARDS NATURAL SCIENCE ETABLISH
761-0255EA 464 EUR
761-0255
Osteometric board
Opetusmateriaalit Forensics Educational Materials Crime Scene Evidence Learning Activities
The osteometric board features a sturdy fixed head piece and sliding foot piece that re operated with a smooth, fluid motion to allow for precise and accurate measurement of long bones up to 600 mm.
  • Durable, lightweight PP
  • Easy to use and transport
The scale is accurate, easy-to-read and graduated in mm to eliminate the need for conversions in standard regression formulas.