Sketching traffic template set, Ward's®
WARD150222EA 67.8 EUR
WARD150222
Sketching traffic template set, Ward's®
The Traffic Template outlines vehicles, structures, and symbols for accident re-creation. The conversion scale for all templates is 1:48, with a small section of the crime scene template at 1:120.
  • Size: 102×204 mm