Detection pen for counterfeit US banknotes, Ward's®
WARD151860EA 26 EUR
WARD151860
Detection pen for counterfeit US banknotes, Ward's®
The same tool used in banks and retail locations to quickly authenticate a suspicious large denomination banknote, this pen will make pale yellow marks on true US paper currency and brown coloured marks on counterfeit bills.
  • Length: 134 mm