Incremental grids, Ward's®
Toimittaja: WARDS NATURAL SCIENCE ETABLISH
818-0145EA 33.5 EUR
818-0145 818-0144
Incremental grids, Ward's®
Opetusmateriaalit Forensics Educational Materials Crime Scene Evidence Learning Activities
Transparent metric grids with red graphics maximise overlay visibility.
  • Matte finish is non-reflective for photography
  • Each has a pair of 1 mm graduated axes which transcribe 1 cm2 grid windows