Creating a composite sketch using FACES software activity guide
Toimittaja: Ward's
Ward's®
761-0298EA 26.9 EUR
761-0298
Creating a composite sketch using FACES software activity guide
Opetusmateriaalit Forensics Educational Materials Crime Scene Evidence Learning Activities
Practical application of the revolutionary program.
  • Convenient illustration program
Sharpen student's observational and communication skills while they create accurate facial composites in this lab activity. After studying the included 8×10" photographs representing suspects, students take turns being witnesses as they describe the face to a student acting as an officer. Using your FACES 4.0 EDU software, 'officer' students create sketches of the suspect and compare their final result with the pictures provided. For an added element of intrigue and entertainment, you can commission members of your school’s staff to pose as suspects and have students describe and sketch the faces of their teachers. Includes five close-up photographs. Activity requires FACES software, available separately.