Crime scene V: The abandoned shed lab activity
Toimittaja: Ward's
761-0309EA 186 EUR
761-0309
Crime scene V: The abandoned shed lab activity
Opetusmateriaalit Forensics Educational Materials Laboratoriosarjat
Become the crime scene investigator charged with conducting five blood analysis tests to help solve the mystery of the abandoned shed, which seems to be the centre of a lot more than garden tool storage. In this activity police are notified by a mysterious caller with information that has led them to the scene of a horrible crime of murder. To solve the case student conduct ABO and Rh blood typing, Kastle-Meyer, Benzidine, and Barr Body tests to identify the killer.
  • Blood evidence analysis solves the crime
  • Multiple techniques
Toimitustiedot: Set includes materials for three classes of 24 students along with instructions and reference materials in a bound booklet for easy duplication.