Phenolphthalein blood test kit
Toimittaja: Ward's
761-0329EA 42.5 EUR
WARD366134
Phenolphthalein blood test kit
Save time in laboratory experiments using the premixed chemicals and plastic dropper bottles of liquid reagents in this kit. Note: The phenolphthalein test will not distinguish between human and animal blood. Further serology analysis is necessary.
Toimitustiedot: Supplied with materials required for the presumptive determination of real animal blood, and plastic storage case, for 30 tests.