Inkless pocket print kit, Ward's®
WARD366137EA 101 EUR
WARD366137
Inkless pocket print kit, Ward's®
The easiest, cleanest, and quickest way to obtain fingerprints in the field.
  • Convenient to use
  • Mess-free record keeping
Produce sharp, dark prints instantaneously with the unique elements of this portable kit, all bound in a leatherette cover. Simply roll the fingers on the yellow coating pad attached to the case, then roll the fingers on the special chemically–treated paper; no visible residue will remain. Enough solution is included to develop 10 prints on each of the 50 sheets provided, which also allow room for recording case number and personal information on their reverse sides.