AA_US_Magnetic fingerprint kit
Toimittaja: Ward's
WARD366139EA 149 EUR
WARD366139
AA_US_Magnetic fingerprint kit
Uncover latent fingerprints like a professional with the easy to use and duplicate fingerprinting techniques.
  • Includes 90 ml wide-mouth powder jar for convenient use with the magnetic applicator
Contained in a sturdy carrying case (264×117×45 mm).
Toimitustiedot: Includes a magnetic powder applicator, 1 oz./28 g of bi–chromatic magnetic powder, 38 mm×9,14 m roll of lifting tape, and 76×127 mm cards (100), all contained in a sturdy carrying case.