Sets of preprepared slides for comparison for a wide range of crime scene evidence types.
- A Selection of several types of crime scene evidence; includes blood, hair, fibres, human cells and fingerprints
Blood and sperm: Differentiate between species by blood elements or morphological differences; set of smears slides -including human, livestock, pets and crustaceans.
Hair and fibre: Identify structural differences found in fibre and hair evidence; distinguish between animal and human hair found at crime scenes; whole fibres and hairs are mounted on the slides.
Fingerprint types: Identify individual friction ridge patterns- whirls, arches and loops.
Human pathology: Selection of common histological afflictions, cancer tissues to goiter.
Trace evidence: Use the slides to reference the microscopic evidence; slides range from dusts and pollens to toothpaste.
Toimitustiedot: For full details of the contents of each set, please ask for details.