Light meter, DL-204 LUX
Toimittaja: GOnDO Electronic
763-0508EA 423 EUR
763-0508
Light meter, DL-204 LUX
Valomittarit
High precision meter measuring intensities of illumination in Lux or Footcandles.
  • Data hold
  • Max. and Zero adjustment
  • Compact size
Sensor: Silicon photodiode and filter