Sound meter
Toimittaja: GOnDO Electronic
763-0294EA 259 EUR
763-0294
Sound meter
Äänitason mittauslaitteet, äänimittarit
Lightweight pocket model for dBA measurements between 35 to 120 dB. Battery check for 9 V battery and equipped with electronic calibration. Reliable and stable microphone. Accuracy ± 5 dBA.