(4-Acetylphenoxy)acetic acid ≥98%

Varoitus

L00426.22EA 764 EUR
L00426.22 L00426.14
(4-Acetylphenoxy)acetic acid ≥98%
(4-Acetylphenoxy)acetic acid
Kaava: (C₆H₄(C(O)CH₃))OCH₂COOH
MW: 194,19 g/mol
Sulamispiste: 172…174 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00014363
CAS-numero: 1878-81-5
EINECS: 217-524-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR