6-Chloronicotinonitrile

Toimittaja: Molekula

MOLE87085479-5GEA 0 EUR
MOLE87085479-5G
6-Chloronicotinonitrile
6-Chloronicotinonitrile
Kaava: C₆H₃ClN₂
MW: 138,56 g/mol
MDL numero: MFCD00084941
CAS-numero: 33252-28-7
UN: 3439
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR