Antraliini

EDQMD2953000EA 282 EUR
EDQMD2953000
Antraliini
Antraliini
Kaava: C₁₄H₁₀O₃
MW: 226,23 g/mol
Kiehumispiste: 400 °C (1013 hPa)
Sulamispiste: 193,5…195 °C
Tiheys: 1,433 g/cm³ (20 °C)
MDL numero: MFCD00053409
CAS-numero: 1143-38-0
EINECS: 214-538-0
Merck Index: 12,00723

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR