Elohopeabromidi -testipaperi for the determination of arsenic, Supelco®
Toimittaja: Merck
SUPL83544-1EAEA 37.9 EUR
SUPL83544-1EA
Elohopeabromidi -testipaperi for the determination of arsenic, Supelco®
Determination of arsenic in grape must and wine, determination of arsine in air. In the latter case the test paper should first be moistened with acetic acid anhydride.