Burettes, straight stopcock, Assistent®
Toimittaja: Hecht
Assistent®
HECH40799050EA 72.6 EUR
HECH40799050 612-3767 612-3766
Burettes, straight stopcock, Assistent®
Byretit
Clear glass burette with graduations.
  • Borosilicate glass 3.3
  • Class B
  • NS stopcock with PTFE key