Suojapuku, laboratorio, MS 356, PP
Toimittaja: Honeywell Safety Products
113-0410EA 237 EUR
DALL4571433/L DALL4571433/M DALL4571433/XXL DALL4571433/XL
Suojapuku, laboratorio, MS 356, PP
Disposable garment made from PP with hood for protection from chemicals. Elasticated at wrists, ankles and back. Zip with double placket for protection (top and underside) extending to chin.
  • No seams on arms and shoulders for extra protection
  • Multi-layer fleece, effective in terms of protection and comfort
  • Loose fit for extra comfort and durability
  • Quick and easy to put on
Weight: 55 g
Väri: valkoinen
Sertifikaatit: Type 5/6, EN 340, EN 1149-1, Cat. III