Viljelyastiat, viljelysäiliöt, Techne®
Toimittaja: Cole Parmer
Techne®
734-0882EA 543 EUR
734-0882 442-0706 734-0881 734-0880
Viljelyastiat, viljelysäiliöt, Techne®
Kolvit ja erlenmeyerit Viljelyastiat
Culture flasks, borosilicate glass, siliconised, for use with the MCS platform biological stirrers, ideal for suspension cell culturing with reduced cell attachment.
  • Incorporate a unique base design which, together with the bulb-ended stirrer, ensures that cultures are lifted into suspension at the lowest possible speeds preventing cell damage
  • Can be sealed for use with pathogenic materials
  • Culture vessels are siliconised to reduce the possibility of cells attaching to and growing on the surfaces