Stoppers for infusion and transfusion flasks
Toimittaja: Saint-Gobain
VERN443302EA 300 EUR
VERN443302
Stoppers for infusion and transfusion flasks
Butyl rubber stopper, halogenated in red.
  • Very good physical and chemical properties
  • Low reabsorption of moisture
  • Suitable for lyophilisates