These earth friendly thermometers feature green, non toxic, biodegradable, enviro safe liquid against lead-free, white back glass. Ideal for use in chemical/petrochemical laboratories, universities, scientific institutions, food and beverage processing, pharmaceutical and medical industries, dairies, breweries and distilleries.
- Enviro safe täyte
- Lyijytön lasi
- Yksittäiset sarjanumerot
- Tarkkuus ilman mitään elohopean aiheuttamia vaaroja
Sertifikaatit: Standardien Envirokleen™, NIST mukainen.
Toimitustiedot: Includes a multi-language (English, French, German, Italian, Portuguese and Spanish) statement of accuracy and traceability.