VWR® ND10, ruuvikaulapullot
Toimittaja: Avantor
548-3187EA 655 EUR
VWRI548-3187 VWRI548-0385
VWR® ND10, ruuvikaulapullot
These vials are intended for use on instruments from the following manufacturers: Jasco, PerkinElmer, Shimadzu, Varian, Waters®, more.
  • Wide opening enables easy filling with viscous materials
  • Any combination of 1,5 ml screw neck vial 10-425 with one of the 10 mm PP screw seals can be ordered as a 2 in 1 kit
  • Closed top seals and replacement septa are available
  • Broad range of micro-inserts