Puhdastilan pyyhkeet, esikyllästetty, steriilit, PROSAT® Sterile™
Toimittaja: Contec
PROSAT® Sterile™
115-0678EA 1020 EUR
115-0678 115-0781 115-0782
Puhdastilan pyyhkeet, esikyllästetty, steriilit, PROSAT® Sterile™
Pyyhkeet, liinat ja sienet Puhdastilan pyyhkeet Esikyllästetyt puhdastilan pyyhkeet
These non woven wipes are designed for pharmaceutical, biotech, and medical device applications. Choose between economical hydroentangled polyester/cellulose wipes or meltblown polypropylene wipes for improved durability.
  • Designed for sterile environments
  • Presaturated wipes are convenient and safer to use than liquid IPA
  • Compact pouch easily fits into isolators, bio-safety cabinets and glove boxes
Ideal for cleanup following BD RODAC™ plate sampling, as well as for wiping isolators, cabinets, hoods, and other cleanroom equipment.
Suitable for use in Class 100, ISO 5, M3.5 cleanrooms.
Colour: White
Pakkaus: Wipes are packed in resealable, tamper-evident pouches that preserve product integrity and cleanliness. Pouches are individually lot-numbered for traceability.