Kloorivetyliuos ≥32%, Puriss. p.a., Fluka™

Toimittaja: Honeywell Chemicals
Vaara

30720-2.5LEA 64.68 EUR
30720-2.5L
Kloorivetyliuos ≥32%, Puriss. p.a., Fluka™
Kloorivetyliuos
Kaava: HCl
MW: 36,46 g/mol
Kiehumispiste: 42 °C
Sulamispiste: -35 °C
Tiheys: 1,16 g/cm³ (20 °C)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00011324
CAS-numero: 7647-01-0
UN: 1789
ADR: 8,II

Order Now

Testitulosten erittely

Assay Min. 32 %
Residue on ignition (SO₄) Max. 0.0005 %
Ammonium (NH₄) Max. 0.0001 %
Aluminium (Al) Max. 0.000005 %
Arsenic (As) Max. 0.000001 %
Barium (Ba) Max. 0.000002 %
Beryllium (Be) Max. 0.000002 %
Bismuth (Bi) Max. 0.00001 %
Calcium (Ca) Max. 0.00005 %
Cadmium (Cd) Max. 0.000001 %
Cobalt (Co) Max. 0.000001 %
Chromium (Cr) Max. 0.000002 %
Copper (Cu) Max. 0.000002 %
Iron (Fe) Max. 0.00002 %
Germanium (Ge) Max. 0.000005 %
Potassium (K) Max. 0.00001 %
Lithium (Li) Max. 0.000001 %
Magnesium (Mg) Max. 0.00001 %
Manganese (Mn) Max. 0.000001 %
Molybdenum (Mo) Max. 0.000002 %
Sodium (Na) Max. 0.00005 %
Nickel (Ni) Max. 0.000002 %
Lead (Pb) Max. 0.000002 %
Strontium (Sr) Max. 0.000001 %
Titanium (Ti) Max. 0.00001 %
Thallium (Tl) Max. 0.000005 %
Vanadium (V) Max. 0.000001 %
Zinc (Zn) Max. 0.000005 %
Zirconium (Zr) Max. 0.00001 %
Bromide (Br) Max. 0.005 %
Free chlorine (Cl) Max. 0.00005 %
Phosphate (PO₄) Max. 0.00005 %
Sulfate (SO₄) Max. 0.0001 %
Sulfite (SO₃) Max. 0.0001 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR