2-Hydroxyacetophenone 97%

Toimittaja: Thermo Scientific
Varoitus

ACRO411900250EA 170 EUR
ACRO411900250
2-Hydroxyacetophenone 97%
2-Hydroxyacetophenone
Kaava: C₈H₈O₂
MW: 136,15 g/mol
Kiehumispiste: 118…120 °C (11 mmHg)
Sulamispiste: 83…88 °C
MDL numero: MFCD00041829
CAS-numero: 582-24-1
EINECS: 209-480-8

Order Now

Testitulosten erittely

Appearance Light yellow to orange Powder
Infrared spectrum Conforms
Melting point 83 to 88 °C
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR