Hydrokortisoni

EDQMH1300000EA 282 EUR
EDQMH1300000
Hydrokortisoni
Hydrokortisoni
Kaava: C₂₁H₃₀O₅
MW: 362,47 g/mol
Kiehumispiste: 567 °C (1013 hPa)
Sulamispiste: 202…204 °C
Tiheys: 1,4 g/cm³ (20 °C)
MDL numero: MFCD00011654
CAS-numero: 50-23-7
EINECS: 200-020-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR