2-Propanoli 99.8% for electronic use (MOS) residue-free
Toimittaja: Thermo Scientific
|
Vaara
|
Synonyms:
Isopropyylialkoholi
ACRO327930025EA
114
EUR
ACRO327930025
ACRO327930010
2-Propanoli 99.8% for electronic use (MOS) residue-free
2-Propanoli
Kaava:
(CH₃)₂CHOH MW: 60,1 g/mol Kiehumispiste: 81…83 °C (760 mmHg) Sulamispiste: –89,5 °C Tiheys: 0,785 g/cm³ Leimahduspiste: 12 °C Storage Temperature: Huoneenlämpö |
MDL numero:
MFCD00011674 CAS-numero: 67-63-0 EINECS: 200-661-7 UN: 1219 ADR: 3,II REACH: 01-2119457558-25 Merck Index: 13,05228 |
![]() |
Testitulosten erittely
Appearance | Clear liquid |
Water | ≤0.05 % (Coulometric) |
GC | ≥99.75 % |
Color scale | ≤10 APHA |
Residue after evaporation | ≤0.0005 % |
Acidity | ≤0.0002 meq/g |
Chloride (Cl) | ≤2 ppm |
Arsenic (As) | ≤10 ppb |
Copper (Cu) | ≤100 ppb |
Iron (Fe) | ≤100 ppb |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...