Keräysastiat, Chemware®
Toimittaja: Saint-Gobain
VERND1069102EA 154 EUR
VERND1069102
Keräysastiat, Chemware®
Resistant to most chemicals, the Chemware® dipper is ideal for ultra-pure applications and use with aggressive media.
  • Non contaminating
  • Superior chemical resistance
  • Lowest concentration of metal ions of any fluoropolymer
Handle design allows dipper to hook over edge of vessel, eliminating dripping of corrosive fluids onto surrounding surface. Use with containers that have at least an 80 mm opening.