Lisävarusteet, Torque wrench fittings for PFA closures, Nalgene®
Toimittaja: Thermo Fisher Scientific
Nalgene®
NALG2195-3020EA 193 EUR
NALG2195-3020
Lisävarusteet, Torque wrench fittings for PFA closures, Nalgene®
Epoxy, specifically designed for Nalgene perfluoralkoxy (PFA) closures.
Proper torquing of Nalgene closures is critical to leakproof sealing.