Mittalasit, PFA, Chemware®
Toimittaja: Saint-Gobain
Chemware®
VERND1069606EA 2370 EUR
VERND1069606
Mittalasit, PFA, Chemware®
Mittalasit Measuring Cylinders
PFA, tall form.
  • Easy-to-read red graduations
  • Smooth pouring, drip-free spout
  • Broad round base for stability
  • Temperature resistant from -196 to 260 °C