Bentsoehappo ≥99.5% ACS
Toimittaja: Thermo Scientific
Vaara
36230.22EA 122 EUR
36230.22 36230.36 36230.14
Bentsoehappo ≥99.5% ACS
Bentsoehappo
Purity: 99.5% min
CAS No.: 65-85-0
Molecular Formula: C7H6O2
Formula Weight: 122.12
Flash Point: 121°(250°F)
Physical Form: Crystalline
Density: 1.32
Appearance: White
Odour: Weak, characteristic
pH: 2.5-3.5
Melting Point: 121-124°
Boiling Point: 249°
Beilstein Reference: 636131
Merck Reference: 14,1091
MDL No.: MFCD00002398
Kaava: C₇H₆O₂
MW: 122,12 g/mol
Kiehumispiste: 249 °C
Sulamispiste: 121…124 °C
Tiheys: 1,32
Leimahduspiste: 121 °C (250 °F)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00002398
CAS-numero: 65-85-0
EINECS: 200-618-2
REACH: 01-2119455536-33
Merck Index: 13,01092
https://fi.vwr.com/stibo/web/std.lang.all/81/11/748111.jpg