Letku, polypropeeni, Nalgene®
Toimittaja: Thermo Scientific
NALG8020-0370EA 114 EUR
NALG8020-0370
Letku, polypropeeni, Nalgene®
PP, translucent
  • Very good chemical resistance
  • Very good temperature resistance
This polypropylene tubing with durometer of 75 (Shore D) is unaffected by most solvents at ambient temperatures.
Sertifikaatit: Food-grade materials comply with 21 CFR 177.1520 for food packaging