L-Threonine

EDQMT1340000EA 282 EUR
EDQMT1340000
L-Threonine
L-Threonine
Kaava: C₄H₉NO₃
MW: 119,12 g/mol
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00064270
CAS-numero: 72-19-5
EINECS: 200-774-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR