2-(N-Ethylanilino)ethanol 96%
Toimittaja: Thermo Scientific
|
Varoitus
|
ACRO325221000EA
106
EUR
ACRO325221000
2-(N-Ethylanilino)ethanol 96%
2-(N-Ethylanilino)ethanol
Kaava:
C₁₀H₁₅NO MW: 165,24 g/mol Kiehumispiste: 276 °C Sulamispiste: 33 °C Leimahduspiste: 120 °C Storage Temperature: Huoneenlämpö |
MDL numero:
MFCD00020575 CAS-numero: 92-50-2 EINECS: 202-160-9 |
Testitulosten erittely
Appearance | Clear colorless to yellow Liquid or solid (May darken on storage) |
Infrared spectrum | Conforms |
Refractive index | 1.5600 to 1.5640 (20°C, 589 nm) |
GC | ≥95 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...