7-Benzyloxyindole

Toimittaja: Molekula

MOLE14077435-1GEA 70.3 EUR
MOLE14077435-1G
7-Benzyloxyindole
7-Benzyloxyindole
Kaava: C₁₅H₁₃NO
MW: 223,27 g/mol
MDL numero: MFCD00037974
CAS-numero: 20289-27-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR