1-Butyl-2,3-dimethylimidazoliumbis(trifluoromethylsulphonyl)imide

Toimittaja: Merck
Vaara


M-Clarity™ quality level = MQ200

4.90288.0100EA 869 EUR
4.90288.0100
1-Butyl-2,3-dimethylimidazoliumbis(trifluoromethylsulphonyl)imide
1-Butyl-2,3-dimethylimidazoliumbis(trifluoromethylsulphonyl)imide
Kaava: C₁₁H₁₇F₆N₃O₄S₂
MW: 433,4 g/mol
Storage Temperature: Huoneenlämpö
MDL numero: MFCD10566879
CAS-numero: 350493-08-2
UN: 2810
ADR: 6.1,III

Order Now

Testitulosten erittely

Solubility (20 °C) insoluble
Molar Mass 433.39 g/mol
Density 1.437 g/cm3 (20 °C)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR