Multi purpose beakers, ratiolab®
Toimittaja: Ratiolab
RATI5921171EA 118 EUR
RATI5921171
Multi purpose beakers, ratiolab®
Glass-clear, PS beakers with graduation.
  • Self-adhesive film cover, leakproof and easily labelled
  • Comfortable sampling, the film cover can be pierced with pipette tips
Pakkaus: Multi purpose beakers, 50 are stacked in PE-bags for use with the beaker dispenser.