Tropine

EDQMT2580300EA 282 EUR
EDQMT2580300
Tropine
Tropine
Kaava: C₈H₁₅NO
MW: 141,21 g/mol
MDL numero: MFCD00180667
CAS-numero: 120-29-6
EINECS: 204-384-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR