Copper(II) trifluoromethanesulphonate

Toimittaja: Molekula

MOLE11742172-5GEA 0 EUR
MOLE11742172-5G
Copper(II) trifluoromethanesulphonate
Copper(II) trifluoromethanesulphonate
Kaava: C₂CuF₆O₆S₂
MW: 361,69 g/mol
MDL numero: MFCD00077492
CAS-numero: 34946-82-2
EINECS: 252-300-8
UN: 3261
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR