1,4-Dioksaani, vedetön ≥99.8%
Toimittaja: Thermo Scientific
Vaara
Synonyms: p-Dioksaani
Packaged under argon in resealable ChemSeal™ bottles.
43821.AEEA 154 EUR
43821.AE 43821.M1 43821.K2
1,4-Dioksaani, vedetön ≥99.8%
1,4-Dioksaani
Purity: 99.8%
CAS No.: 123-91-1
Molecular Formula: C4H8O2
Formula Weight: 88.11
Flash Point: 12°(54°F)
Physical Form: Liquid
Density: 1.034
Refractive Index: 1.4220
Sensitivity: Hygroscopic
Odour: Ether-like
Melting Point: 11.8°
Boiling Point: 100-102°
Beilstein Reference: 102551
Merck Reference: 14,3300
UN No.: 1165
MDL No.: MFCD00006571
Kaava: C₄H₈O₂
MW: 88,11 g/mol
Kiehumispiste: 100…102 °C
Sulamispiste: 11,8 °C
Tiheys: 1,034
Leimahduspiste: 12 °C (54 °F)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00006571
CAS-numero: 123-91-1
EINECS: 204-661-8
UN: 1165
ADR: 3,II
Merck Index: 13,03330
https://fi.vwr.com/stibo/web/std.lang.all/38/27/773827.jpg