Natriumhydroksidi, tested according to Ph. Eur., Fluka™
Toimittaja: Honeywell Chemicals
|
Vaara
|
71694-5KGEA
135.67
EUR
71694-5KG
Natriumhydroksidi, tested according to Ph. Eur., Fluka™
Natriumhydroksidi
Kaava:
NaOH MW: 40 g/mol Kiehumispiste: 1,390 °C Sulamispiste: 319 °C Tiheys: 2,13 g/cm³ (20 °C) Storage Temperature: Huoneenlämpö |
MDL numero:
MFCD00003548 CAS-numero: 1310-73-2 EINECS: 215-185-5 UN: 1823 ADR: 8,II REACH: 01-2119457892-27 Merck Index: 14,08627 |
Testitulosten erittely
Assay | 97.0 - 100.5 % |
pH | Min. 11.0 |
Appearance of the solution | complying |
Iron (Fe) | Max. 10 ppm |
Heavy metals (as Pb) | Max. 20 ppm |
Assay of Na₂CO₃ | Max. 2 % |
Chloride (Cl) | Max. 100 ppm |
Sulfate (SO₄) | Max. 100 ppm |
Residual solvents | complying |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...