Palladium 3% on activated carbon for use as a catalyst, unreduced
Toimittaja: Thermo Scientific
|
Varoitus
|
ACRO217870100EA
116
EUR
ACRO217870100
Palladium 3% on activated carbon for use as a catalyst, unreduced
Palladium
Kaava:
Pd MW: 106,420 g/mol Storage Temperature: Huoneenlämpö |
MDL numero:
MFCD00011167 CAS-numero: 7440-05-3 EINECS: 231-115-6 UN: 3178 ADR: 4.1,III |
Testitulosten erittely
Appearance | Black Powder |
Assay | 3 % (Pd) |
Additional info | typical properties: |
Additional info | specific surface area (m²/g): 1000 |
Additional info | total pore volume (ml/g): 1.00 |
Additional info | typical particle size distribution: |
Additional info | D50: 20 um |
Additional info |
|
Additional info |
|
Additional info |
|
Additional info |
|
Additional info |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...