Digital thermometer, pocket size, Traceable®
620-2010EA 141 EUR
VWRI620-2010
Digital thermometer, pocket size, Traceable®
This small, lightweight (113 g) and shockproof thermometer is designed for simple operation in plant and field use.
  • Two probes for simultaneous monitoring of two events
  • Three-position switch indicates probe 1, probe 2, or Off
  • Expanded range, fast response, and one-switch operation
W×D×H: 76×25,4×127 mm
Sertifikaatit: Compliant with ISO/IEC 17025, ANSI/NCSL Z540-1 and 9001.
Toimitustiedot: Supplied with two K-type probes with 120 mm cables, a 9 volt battery and a Traceable calibration certificate according to standards from NIST.
Varoitus: Values are valid for included K-type probes. The thermometer can measure temperatures from –40 to +1200 °C with suitable probes that need to be ordered separately.